missing translation for 'onlineSavingsMsg'
Learn More

Cottonseed oil, pure

CAS: 8001-29-4

$76.21 - $274.69

Chemical Identifiers

CAS 8001-29-4
MDL Number MFCD00130872
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC177115000
SDS View Documents
Thermo Scientific Chemicals
177115000
500 mL
Each for $76.21
Only null left
 
AC177110025
SDS View Documents
Thermo Scientific Chemicals
177110025
2.5 L
Each for $274.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8001-29-4
MFCD00130872

Specifications

0.5mg KOH/g max.
Yellow
>112°C
Glass bottle
MFCD00130872
185 to 198mg KOH/g
10,2535
Pure
Cottonseed oil
8001-29-4
0.9100g/mL
Authentic
1.4720 to 1.4730
500 mL
0.91
Solubility in water: .. Other solubilities: soluble in ether,benzene,chloroform and carbon,disulfide,slightly soluble in alcohol
Oil
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 232-280-7

RTECSNumber : GN2815000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only