missing translation for 'onlineSavingsMsg'
Learn More

Innovatek Cotton Tip Applicators

Catalog No. 2980106
Change view
Click to view available options
Length (English) Handle:
3 in.
6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Handle
2980106 6 in.
2980105 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2980106 Supplier Innovatek Supplier No. 420240200
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek Cotton Tip Applicators available with 3, or 6 in. wooden handles

  • Disposable, cotton tip applicators can be used for a variety of applications
  • Choose from a short (3 in.), or long (6 in.) wooden handle depending on uses

Specifications

Color White, Brown
Product Type Disposable Cotton Swab
Tip Material Cotton
Tip Type Rounded
Length (English) Handle 6 in.
Quantity 1000 Swabs/Box