missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cotinine, 98%

Catalog No. p-7045758
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11873MD 250 mg
AAL1187303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11873MD Supplier Thermo Scientific Chemicals Supplier No. L11873MD

CAS: 486-56-6 | C10H12N2O | 176.219 g/mol

(-)-Cotinine is used to activate a subpopulation of α3/ α6β2 nAChRs in monkey striatum. It binds nicotinic- and muscarinic-type acetylcholine receptors with minimal receptor desensitization and demonstrates antipsychotic drug-like properties in behavioral models, neuroprotective properties in neurodegenerative disease models, and enhances attention in a delayed matching-to-sample task.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible or difficult to mix in water. Soluble in dimethyl sulfoxide (100 mM), ethanol (50 mg/ml, yielding a clear, faint yellow to yellow solution), methanol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.

Chemical Identifiers

CAS 486-56-6
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
MDL Number MFCD00077696
InChI Key UIKROCXWUNQSPJ-VIFPVBQESA-N
Synonym cotinine, --cotinine, cotinina, cotininum, cotinine-, s-cotinine, s---cotinine, unii-k5161x06ll, s-1-methyl-5-3-pyridinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-methyl-5-3-pyridinyl-, 5s
PubChem CID 854019
ChEBI CHEBI:68641
IUPAC Name (5S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
SMILES CN1C(CCC1=O)C2=CN=CC=C2

Specifications

Melting Point 34°C to 39°C
Boiling Point 145°C to 147°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 250 mg
Beilstein 83099
Merck Index 14,2553
Solubility Information Not miscible or difficult to mix in water. Soluble in dimethyl sulfoxide (100 mM),ethanol (50mg/ml,yielding a clear,faint yellow to yellow solution),methanol and chloroform.
Optical Rotation −20° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (-)-Cotinine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-634-9
RTECSNumber GN1925500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only