missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polypropylene PCR Tubes with Attached Cap
GREENER_CHOICE

Catalog No. 07200493
Change view
Click to view available options
Color:
Clear
Closure Type:
Domed Cap
Flat Cap
Volume (Metric):
0.2 mL
0.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Volume (Metric)
07200493 Clear Domed Cap 0.2 mL
07200252 Clear Flat Cap 0.5 mL
07200681 Clear Flat Cap 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07200493 Supplier Corning™ Supplier No. 6531

Manufactured from thin-walled polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Corning™ PCR Tubes withstand forces up to 10,000 x g.

Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.

Specifications

Certifications/Compliance RNase, DNase-free
Closure Color Clear
Color Clear
Volume (Metric) 0.2 mL
Quantity 96/Pk.
Closure Material Polypropylene
For Use With (Equipment) Heat blocks for fast heat transfer
Shape Conical bottom
Sterility Non-sterile
Material Polypropylene
Max. Force 10,000 x g
Closure Type Domed Cap
Autoclavable Autoclavable
Format Tube
Product Type Thin Wall PCR Tube
Show More Show Less