missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Supplier: Corning™ 4401
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.
Chemical Identifiers
| Autoclavable | |
| Translucent | |
| Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
| 96 |
| Polypropylene | |
| 1 mL | |
| 0.033 oz. |
Specifications
| Tube | |
| Bulk | |
| Non-sterile | |
| 960/Pk. | |
| Translucent | |
| Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
| 96 |
| Microplate Cluster Tube | |
| Autoclavable | |
| Polypropylene | |
| 960/Cs. | |
| 1 mL | |
| 0.033 oz. |