missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Clear Multiple Well Plates
GREENER_CHOICE

Catalog No. 07201589
Change view
Click to view available options
Culture Area:
1.9 cm2
3.8 cm2
9.5 cm2
Volume (Metric) Well:
3.4 mL
6.9 mL
16.8 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Volume (Metric) Well
07201589 3.8 cm2 6.9 mL
07201590 1.9 cm2 3.4 mL
07201588 9.5 cm2 16.8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07201589 Supplier Corning™ Supplier No. 3737/DUP

Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Specifications

Format Standard
Lid With Lid
Culture Area 3.8 cm2
Array 3 x 4
Binding Type Medium Binding
Color Clear
Material Polystyrene
Volume (Metric) Well 6.9 mL
No. of Wells 12
Volume (Metric) Working 0.76 to 1.14 mL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 100/Cs.
No. per Pack 5/Pk.
Show More Show Less