Learn More
Corning™ Aspirator
Catalog No. 07200564
Change view
Click to view available options
Type:
Aspirator
Catalog No. Type
07200564 Aspirator
1 options
Catalog No. 07200564 Supplier Corning™ Supplier No. C4930
  • ONLINE EXCLUSIVE PRICE
    $917.81
    $570.50 / Case
    Save $347.31
    38.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Handpiece, grommet for attachment of accessories, and a single-channel adapter for use with disposable pipetter tips.
Ideal for removal of supernatants - an alternative to glass Pasteur pipets
The Corning™ Aspirator is a device using vacuum to safely and aseptically dispose of supernatant from a variety of laboratory vessels. The aspirator includes a hand piece to which disposable pipet tips are attached during use, a grommet for accessory attachment and single-channel adapter for disposable pipet tips.
An 8-channel adapter is available for working in 96 well plates, see No. 4931. Refer to the accessories tab below.
Specifications
Autoclavable Autoclavable
Product Line Costar
Type Aspirator
For Use With (Application) Safe removal/disposal of supernatants from a wide range of laboratory vessels
For Use With (Equipment) Disposable pipette tip, Polyethylene and Polystyrene aspiration pipette, or Pasteur pipette
Includes Handpiece, grommet for accessory attachment and single channel adapter for disposable pipette tip