missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Half-Area Plates
GREENER_CHOICE

Catalog No. 0720093
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
0720093 Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0720093 Supplier Corning™ Supplier No. 3696

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Saves on valuable reagents by reducing the amount of reagent required per well, while retaining the ability to be read in standard microplate readers. Corning™ 96-Well Half Area Microplates are an economical solution for all of your assay diagnostic needs and are available in different colors and treatments.

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Barcode Alphanumeric
Format Half-Area
Lid With Lid
Culture Area 0.16 cm2
Array 8 x 12
Packaging Individually Wrapped
Binding Type TC-Treated
Color Clear
For Use With (Equipment) Plate readers
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Enhance luminescent signals and have low background luminescence and fluorescence
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear
Show More Show Less