missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate

Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Half-Area |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Not Treated |
| Color | Clear |
| For Use With (Equipment) | Plate readers |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 25 to 125 μL |
| Show More |