missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Catalog No. 07200334
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200334 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200334 Supplier Corning™ Supplier No. 3697

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Barcode Alphanumeric
Format Half-Area
Lid With Lid
Culture Area 0.16 cm2
Array 8 x 12
Packaging 20/Pk., 100/Cs.
Binding Type TC-Treated
Color Clear
Product Type Half Area
For Use With (Equipment) Plate readers
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Enhance luminescent signals and have low background luminescence and fluorescence
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.
Show More Show Less