missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Clear Polystyrene 96-Well Microplates
GREENER_CHOICE

Catalog No. 0720035
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
0720035 Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0720035 Supplier Corning™ Supplier No. C3590
  • ONLINE EXCLUSIVE PRICE
    $1,713.59
    $576.22 / Case of 100 Each

    Save $1,137.37

    66.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Binding Property High Binding
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Bind
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment High Binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and.or hydrophobic regions
No. per Case 100/Cs.
No. per Pack 1/Pk.
Quantity 100/Cs.
Show More Show Less