missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Description
- Minimal well-to-well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 320μL to 360μL
- Working volume: 75μL to 200μL
- Choice of flat, round or V-shaped bottom
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Specifications
Specifications
| Lid | With Lid |
| Array | 8 x 12 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | Attachment and growth of anchorage-dependent cells |
| Show More |