missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Assay Blocks

Catalog No. 07200700
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Well Shape:
Conical Bottom (V-well)
Round
Round, Conical Bottom (V-well)
No. of Wells:
96
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Well Well Shape No. of Wells
07200700 Sterile 2000 μL Conical Bottom (V-well) 96
07200701 Non-sterile 2000 μL Conical Bottom (V-well) 96
07200724 Non-sterile 500 μL Round, Conical Bottom (V-well) 96
09761116A Non-sterile 1000 μL Round 96
09761116 Sterile 1000 μL Round 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07200700 Supplier Corning™ Supplier No. C3960
Get Up to $2,500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Certifications

Certified DNase- and RNase-free

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Not Treated
Color Clear
Product Type 96-well Assay Blocks
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
For Use With (Application) For compound storage, assay and cell-growth application
No. per Case 25/Cs.
No. per Pack 5/Pk.
Quantity 25/Cs.
Show More Show Less