missing translation for 'onlineSavingsMsg'
Learn More

BlueMed COSMIC Shoe Covers

Numéro de catalogue. 18009106
Change view
Click to view available options
Size:
Universal
X-Large
Packaging Type:
120/Easy Open Bags, 2 Bags/Cs.
150/Dispenser Boxes, 2 Boxes/Cs.
150/Easy Open Bag, 2 Bags/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size Packaging Type
18009106 X-Large 120/Easy Open Bags, 2 Bags/Cs.
18009104 Universal 150/Easy Open Bag, 2 Bags/Cs.
18009105 Universal 150/Dispenser Boxes, 2 Boxes/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 18009106 Fournisseur BlueMed Code fournisseur SHS4501C
Il en reste null

Shoe covers are made with a multi-layer fabric to provide water resistance and anti-skid properties and are ideal for labs and cleanroom environments.

  • Durable protection
  • Strong polypropylene and polyethylene material
  • Fluid resistant and wear resistant
  • Anti-skid
  • Waterproof
  • Low linting, suitable for cleanrooms up to level ISO 5
  • Seamless sole and sides
  • Disposable
  • Available in Universal and XL sizes
  • Made in Canada

Spécifications

Size X-Large
Material Polyethylene, Polypropylene, Polyethylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-skid
Seam Type Ultrasonically Welded
For Use With (Application) GMP or Research Facilities, Pharmaceutical Production, Labs, Cleanrooms
Packaging Quantity 240/Cs.
Packaging Type 120/Easy Open Bags, 2 Bags/Cs.
Length (English) 18.5 in.
Length (Metric) 47 cm
Height (English) 6.99 in.
Height (Metric) 17.75 cm
Weight (Metric) 60 gsm
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100 (ISO 5)
Afficher plus de résultats Afficher moins de résultats