missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products GL603C5 Series Micro-Foam Nitrile-Coated HPPE Cut-Resistant Gloves, Black

Catalog No. p-7182087
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
Catalog No. Size Glove Cuff Color
19555074 Small Red
19555073 Medium Yellow
19555072 Large Brown
19555076 X-Large Black
19555075 2X-Large Blue
5 options

Catalog No. 19555074

Supplier: Pyramex Safety Products GL603C5S

Only null left

MicroFoam nitrile coating acts like a sponge to help increase gripping power, while the HPPE shell provides ANSI Level A4 Cut Protection

  • 13-gauge HPPE shell is resistant to abrasions, cuts, and tears
  • MicroFoam coating enhances grip capabilities
  • Dark color hides dirt and stains for extended useful life

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4X43DX
For Use With (Application) Assembly line, furniture manufacturing, HVAC, aerospace and construction
Glove Cuff Color Red
Glove Cuff Style Continuous Knit
Model GL603C5S
Puncture Rating ASTM F2878 LEVEL 4
Series GL603C5
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov