Learn More
Eisco™ Corundum Mineral Specimen
For study of mineral hardness and crystal habit - EISCO Corundum Mineral Specimen is used in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements.
$156.50
Specifications
Product Type Corundum Mineral Study Specimen
For Use With (Application) Study of mineral hardness and crystal habit
Packaging Clear labeled and barcoded bag
Specimen Size Approx. 1 in.
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S24668
View Documents Promotion Details
Eisco™
ESNG0008PK12
12/Pk.
Pack of 12 Each for $156.50
Only null left
Description
Description
  • For exercises designed to distinguish between crystal habit and cleavage
  • Comes in clear labeled and barcoded bag for ideal storage
  • Average specimen size: 2-3cm
  • Has Mohs scale hardness of 9, making it one of the hardest minerals on planet
  • For use in mineral hardness exercises and lessons in differentiating between cleavage and crystal habit
Specifications
Specifications
Corundum Mineral Study Specimen
Study of mineral hardness and crystal habit
Clear labeled and barcoded bag
Approx. 1 in.