missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 96-Well, Ultra-Low Binding, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07200603
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200603 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200603 Supplier Corning™ Supplier No. C3474
Get Up to $2,500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Color: Clear; Sterility: Sterile; Lid: With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Barcode No Barcode
Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Ultra-Low Attachment
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Ultra-low Binding
Total Volume (Metric) Well 360 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Maintaining cells in a suspended, unattached state
Includes Condensation rings
No. per Case 24/Cs.
No. per Pack 1/Pk.
Quantity 24/Cs.
Diameter (Metric) Well 0.64 cm
Show More Show Less