Learn More
Corning™ Tubing Sets and Connectors for CellCube™
Description
- Closed System Cell Cube
- Helps reduce the risk of adventitious contamination
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
Specifications
| Certifications/Compliance | EN ISO 13485, FDA CFR 21 Part 820, cGMP, BSE/TSE (EMA/410/01), Non-Pyrogenic, USP Class VI Tested, Radiation Sterilized (ANSI/AAMI/ISO 11137), SAL 10⁻⁶ |
| For Use With (Equipment) | Corning CellCube System |
| Includes | Medium Filter, 2 Aseptic Connectors |
| No. per Case | 4/Cs. |
| No. per Pack | 1/Pk. |
| Product Type | T-Tubing Manifold |
| Sterility | Sterile |
| Quantity | 1 Ea. |