missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Thermowell™ GOLD 384-Well Polypropylene PCR Microplates
GREENER_CHOICE

Catalog No. 07201220
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07201220 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201220 Supplier Corning™ Supplier No. 3757

Fit automated equipment as well as the original manufacturers' plates

Corning Thermowell GOLD 384-well PCR microplates feature exceptional dimensional stability following thermal cycling, and are fully compatible with automation, commonly used thermal cyclers, and Applied Biosystems™ sequencing adapters (see compatibility table).

Order Info

Packaged in packs of 10.

Certifications

Certified DNase- and RNase-free.

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
Well Shape Round, Conical Bottom (V-well)
For Use With (Application) For automation
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Clear
Show More Show Less