Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Supplier:  Corning™ 6511
 View more versions of this product
Catalog No. 07200245
Only null left
Description
Description
  • Low protein-binding surface prevents loss of sample
  • For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
  • Withstands temperatures to 135°C without warping
  • Various well shapes fit popular thermal cyclers
  • Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
  • Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
Specifications
Chemical Identifiers
Standard
8 x 12
Polycarbonate
Non-sterile
PCR
1/Pk.
Without Lid
Clear
96
Round
25/Cs.
Clear
Specifications
Standard
8 x 12
Clear
MJ Research PTC-100-96V, PTC-200 DNA EngineTM, Biometra Uno - ThermoblockerTM,Coy Corporation Temp Cycler II, Corbett Research FTS-960, Hybaid OmniGene withMicroblock, Quatro BioSystems T-C-40
96
Round
25/Cs.
Clear
Without Lid
Medium Binding
PCR Microplate, Model M
Polycarbonate
Non-sterile
PCR
1/Pk.
Product Suggestions
Product Suggestions