missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Stripwell™ Microplates
GREENER_CHOICE

Catalog No. 07200595
Change view
Click to view available options
Surface Treatment:
High Binding
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
07200595 High Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200595 Supplier Corning™ Supplier No. 3923

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.

Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.

  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.

Specifications

Binding Property High Binding
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color White
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment High Binding
Total Volume (Metric) Well 360 μL
Well Shape Round
For Use With (Application) In-vitro diagnostic assays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 100/Cs.
Show More Show Less