missing translation for 'onlineSavingsMsg'
Learn More

Corning™ SF Medium

Catalog No. MT40102CV
Change view
Click to view available options
Without Additives:
No Phenol Red
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Without Additives
MT40102CV No Phenol Red
MT40101CV None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT40102CV Supplier Corning™ Supplier No. 40102CV

Based on a proprietary combination of classical media formulations

SF Medium is based on a proprietary classical media formulation. This medium contains a mixture of select trace elements and high molecular weight carbohydrates, extra vitamins, animal-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L). SF Medium supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures. This formulation contains L-glutamine.

Specifications

Product Type SF Medium
Sterility Sterile
With Additives Glutamine
Without Additives No Phenol Red
Quantity 6 x 500 mL
pH 7.2 to 7.6
Content And Storage 2° to 8°C