Learn More
Corning™ Sealing Mat for 96-Well Polypropylene Microplates
Catalog No. 07202504
Change view
Click to view available options
Plate Compatibility:
96-well Polypropylene Plate
Catalog No. Plate Compatibility
07202504 96-well Polypropylene Plate
1 options
Catalog No. 07202504 Supplier Corning™ Supplier No. 3346
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Firm seal prevents evaporation and gas exchange
  • Precise fit for 96 well Expanded Volume Polypropylene Microplate (Item #’s 3343 and 3344)
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Firm seal prevents evaporation and gas exchange
  • Ideal for use in long-term, low temperature compound storage
Specifications
Sterility Non-sterile
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Product Type Sealing Mat
Plate Compatibility 96-well Polypropylene Plate