missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Roller Bottle Caps
GREENER_CHOICE

Catalog No. 07200713
Change view
Click to view available options
Thread Size (Metric):
31 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
07200713 31 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200713 Supplier Corning™ Supplier No. 431132

Easy-grip polyethylene

  • Individually wrapped
  • Sterile

Specifications

Color Red
Material Polyethylene
No. per Case 300/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Thread Size (Metric) 31 mm
Neck Finish 31 mm
For Use With (Equipment) Roller bottle