Learn More
Corning™ Reusable Phenolic Screw Caps With Rubber Liners
Specially formulated phenolic resin resists the effects of temperature and steam in autoclaving
$380.21 - $1,261.16
Specifications
| Material | Phenolic Resin |
|---|---|
| No. per Case | 144/Cs. |
| Closure Color | Black |
| Closure Material | Phenolic |
| Closure Type | Rubber Liner |
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
| Phenolic Resin | |
| Black | |
| Rubber Liner |
| 144/Cs. | |
| Phenolic |