Learn More
Corning™ PYREX™ VISTA™ Class B Volumetric Flasks with Snap Caps
Catalog No. S95185B
Change view
Click to view available options
Capacity (Metric):
50 mL
Catalog No. Capacity (Metric)
S95185B 50 mL
1 options
Catalog No. S95185B
Supplier: Corning™ 7058150/EMD
Only null left
Capacity tolerances are twice those specified for Class A volumetric ware.
  • Large, blue, block letters make inscriptions easy to read
  • Sharp, permanent graduation lines
  • Snap caps are supplied with all sizes
Specifications
Calibration Calibrated To Contain
Closure Color Blue
Closure Material LDPE
Capacity (Metric) 50 mL
Material Glass, Borosilicate Glass
Neck Style Long
Bottom Shape Flat