Learn More
Corning™ PYREX™ Assortment Pack of Griffin Low form Beakers
Catalog No. 361004967
Change view
Click to view available options
Capacity (Metric):
50 mL, 100 mL, 250 mL, 600 mL, and 1000 mL
Catalog No. Capacity (Metric)
361004967 50 mL, 100 mL, 250 mL, 600 mL, and 1000 mL
1 options
Catalog No. 361004967 Supplier Corning™ Supplier No. 1000PACK
Double scale; graduated
A convenience pack containing one each of the most popular sizes of Griffin low form beakers. Designed for the low volume user, a case contains one each of five sizes; 50 mL, 100 mL, 250 mL, 600 mL, and 1 L. Packaged in a partitioned carton for safe transit and storage. Reference: ASTM E-960.
Specifications
Capacity (Metric) 50 mL, 100 mL, 250 mL, 600 mL, and 1000 mL
Material Borosilicate Glass
Type Beaker