Learn More
Corning™ 384-Well Clear Polystyrene Microplates
Catalog No. 0720105
Change view
Click to view available options
Color:
Clear
Catalog No. Color
0720105 Clear
1 options
Catalog No. 0720105 Supplier Corning™ Supplier No. 3702BC
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • Specifications
    Format Standard
    Lid Without Lid
    Array 16 x 24
    Color Clear
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    Includes Barcode labels
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less