missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polypropylene Plates

Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.

$1,067.11 - $1,926.27

Specifications

Format Standard
Lid Without Lid
Color Clear
Material Polypropylene
Surface Treatment Non-treated
View More Specs

Products 7
Catalog Number Mfr. No. Sterility No. of Wells Array Well Shape Price Quantity & Availability  
Catalog Number Mfr. No. Sterility No. of Wells Array Well Shape Price Quantity & Availability  
07200695
View Documents Promotion Details
Corning™
3363
Non-sterile 96 8 x 12 Conical Bottom (V-well)
Case of 100 Each for $1,077.56
Only null left
 
07200696
View Documents Promotion Details
Corning™
3364
Non-sterile 96 8 x 12 Flat Bottom (F-well)
Case of 100 Each for $1,113.94
Only null left
 
07200747
View Documents Promotion Details
Corning™
3656
Sterile 384 16 x 24 Round
Case of 100 Each for $1,926.27
Only null left
 
07200745
View Documents Promotion Details
Corning™
3359
Sterile 96 8 x 12 Round
Case of 100 Each for $1,137.67
Only null left
 
07200748
View Documents Promotion Details
Corning™
3657
Non-sterile 384 16 x 24 Round
Case of 100 Each for $1,884.77
Only null left
 
07200698
View Documents Promotion Details
Corning™
C3357
Sterile 96 8 x 12 Conical Bottom (V-well)
Case of 100 Each for $1,137.53
Only null left
 
07200697
View Documents Promotion Details
Corning™
3365
Non-sterile 96 8 x 12 Round
Case of 100 Each for $1,067.11
Only null left
 
Description

Description

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Order Info

Seal with Storage Mat III (sold separately).

Specifications

Specifications

Standard
Clear
Non-treated
100/Cs.
100/Cs.
Without Lid
Polypropylene
Transport and longterm storage
25/Pk.
Videos
SDS
Documents

Documents