Learn More
Corning™ PYREX™ Polyethylene Standard Taper Stoppers
Improved design fully protects neck in event of accidental tipping
$54.87 - $120.26
Specifications
Description Includes: Three rings on the plug
Color Blue, White
Material HDPE
Product Type Tapered Stopper
Shape Tapered
View More Specs
Products 7
Catalog Number Mfr. No. Stopper Number Price Quantity  
Catalog Number Mfr. No. Stopper Number Price Quantity  
C7624-13
View Documents
Corning™
762413
13
Case of 6 Each for $56.42
Only null left
C7624-27
View Documents
Corning™
762427
27
Case of 6 Each for $96.45
Only null left
C7624-9
View Documents
Corning™
76249
9
Case of 6 Each for $57.77
Only null left
14-629-46A
View Documents
Corning™
762416
16
Case of 6 Each for $65.46
Only null left
C7624-38
View Documents
Corning™
762438
38
Case of 6 Each for $120.26
Only null left
C7624-22
View Documents
Corning™
762422
22
Case of 6 Each for $81.38
Only null left
14-629-46B
View Documents
Corning™
762419
19
Case of 6 Each for $54.87
Only null left
Description
Description
These blue and white stoppers are made with closed bottoms, of linear high-density polyethylene to conform to standard taper stopper dimensions. The three unit rings on the base provide an efficient seal. The improved design fully protects the neck in the event of accidental tipping. These stoppers may be used in place of the conventional standard taper flask stoppers.
Specifications
Specifications
Includes: Three rings on the plug
HDPE
Tapered
Blue, White
Tapered Stopper
Improved design fully protects the neck in the event of accidental tipping
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'