missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix HC
GREENER_CHOICE

Catalog No. p-4906921
Change view
Click to view available options
Without Additives:
No LDEV
No LDEV, No GFR
No LDEV, No Phenol Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Without Additives
08774391 No LDEV, No Phenol Red
08774392 No LDEV, No GFR
CB354248 No LDEV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Catalog No. 08774391 Supplier Corning™ Supplier No. 354262
Only null left
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer

High concentration (HC) formulation is ideal for applications in which a high protein concentration augments the growth of tumors; allows a Matrigel basement membrane matrix plug to maintain its integrity after subcutaneous injection into mice

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications

Concentration High
Product Type Basement Membrane Matrix
Sterility Sterile
Without Additives No LDEV, No Phenol Red
Product Line Matrigel Matrix
Quantity 10 mL
Format Matrigel Matrix High Concentration, Phenol Red-Free, LDEV-Free
Source Engelbreth-Holm-Swarm Mouse Tumor
Content And Storage -20°C