missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Volume Microplate
GREENER_CHOICE

Catalog No. 07202626
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07202626 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07202626 Supplier Corning™ Supplier No. C3542
Only null left

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

Specifications

Format Standard
Lid With Lid
Culture Area 0.03 cm2
Array 16 x 24
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 0.005 to 0.04 mL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) For miniaturized 384-well fluorescent homogeneous assays
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less