missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Low-Volume 384-Well Black Flat Bottom Polystyrene Non-Treated Microplate
GREENER_CHOICE

Catalog No. 07200877
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200877 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200877 Supplier Corning™ Supplier No. 3821
Get Up to $2,500 in Products with Purchase
View all promotions

Ideal for miniaturized 384-well fluorescent homogeneous assays

Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

Specifications

Lid Without Lid
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 0.005 to 0.04 mL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For miniaturized 384-well fluorescent homogeneous assays
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less