missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Hepatocyte Culture Media Kit, 500mL
GREENER_CHOICE

Catalog No. CB30055M
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB30055M 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB30055M Supplier Corning™ Supplier No. C355056

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Corning Hepatocyte Culture Medium (500 mL), EGF (5μg).

500 mL plus EGF supplement. This kit comprises a serum-free, fully-defined medium.

Corning Gentest cryopreserved animal hepatocytes allow researchers to study toxicological and pharmacokinetic properties of new chemical entities (NCEs) in vitro prior to expensive in vivo animal testing. Understanding species differences in metabolism is critical to selecting appropriate preclinical species for toxicity and pharmacokinetic studies. Corning Gentest animal cryohepatocytes are suitable for use in metabolic stability, metabolite profiling, and cytotoxicity studies.

  • Convenience – animal cryopreserved hepatocytes for rat, mouse, dog, and monkey are available as cryopreserved vialed suspension cells; just thaw and perform studies when needed
  • Full characterization – tested for high viability and phase I and II metabolic enzymes

Specifications

Product Type Stem Cell Differentiation Media
Sterility Sterile
For Use With (Application) Hepatocyte Culture
Quantity 1 Kit
Content And Storage 2° to 8°C