missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ GL32 Orange Polypropylene Screw

Description
These are various sizes and types of screw caps for use with storage bottles.
Specifications
Specifications
| Color | Orange |
| Material | Polypropylene |
| No. per Case | 20/Cs. |
| No. per Pack | 20/Pk. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 32 |
| Neck Finish | 39 mm |
| For Use With (Equipment) | Storage bottle |
| Product Type | Plug Seal |