missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Pre-Coated PAMPA Plate System, with Lid
GREENER_CHOICE

Catalog No. 07201237
Change view
Click to view available options
Color:
Transparent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07201237 Transparent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201237 Supplier Corning™ Supplier No. 353015

A 96-well permeable support system with a filter plate pre-coated with structured layers of phospholipids.

  • Stable, pre-coated PAMPA plate is highly reproducible
  • Improved assay predictability over traditional PAMPA
  • Reduced mass retention of compounds
  • Well-suited as a ranking tool for the assessment of compounds with passive transport mechanisms and for predicting in vivo drug permeability
  • Polystyrene construction

Specifications

Certifications/Compliance ANSI standards
Content And Storage -20°C
Array 8 x 12
Pore Size 0.45 μm
Color Transparent
Material Polystyrene
No. of Wells 96
For Use With (Application) Predicting permeability, assessment of compounds
Quantity Cs. of 5
Width (Metric) 127.6 mm
Height (Metric) 21.7 mm
Depth (Metric) 85.3 mm
Show More Show Less