Learn More
Corning™ Flexible Packaging Systems: EVA Collection Bags
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Order Info
The full line of bag types include: collection containers (bottom port and face port designs); tank liners (2D and 3D); cryopreservation bags; cell expansion bags; bags for HYPERStack™ vessels; and rocker cell culture bags. Collection bags, collection containers and media totes are sold individually on a per each basis.
Specifications
Specifications
| Material | EVA |
| Sterility | Sterile, Irradiated |
| No. per Case | 1/Ea. |
| No. per Pack | 1/Pk. |
| Port Detail | MPC connector; M/F luers |
| Capacity (Metric) | 10 L |