Learn More
Falcon™ 96-well Feeder Tray and Lid
Description
Falcon™ 96-well Feeder Tray, with lid. Non-TC-treated Polystyrene cell media tray for feeding cell-based assays. The drop-in baffle for the Feeder Tray mitigates media sloshing and lowers the risk of contamination.
- Gamma irradiated.
- Nonpyrogenic.
Specifications
Specifications
| Lid | With Lid |
| Array | 8 x 12 |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| No. per Case | 5/Cs. |
| No. per Pack | 5/Pk. |
| Quantity | 5/Cs. |