missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ 96-Square Well, Angled-Bottom Plate and Lid

Description
Falcon™ 96 Square Well Angled Bottom Multiwell Insert System, with lid, sterile, 5/case, Non-TC-treated polystyrene, nonpyrogenic. The Falcon 96 Multiwell Insert System is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays.
Automation compatible design Format compatible with most robots and fluid handling instruments Complete sample recovery The Falcon 96 Square Well, Angled-Bottom Plate features an angled bottom for more complete sample utilization Excellent reproducibility One-piece feeder tray enhances consistency in well-to-well monolayer growth Total assay flexibility—ideal for transport studies System can be used with many cell lines including Caco-2, MDCK, and LLC-PK1, for basal to apical or apical to basal measurements of drug transport
Storage: Store at room temperature, away from direct sunlight.
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Includes | Insert, feeder tray and lid |
| No. per Case | 5/Cs. |
| No. per Pack | 5/Pk. |
| Show More |