missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DiIC12(3) Fluorescent Dye

Catalog No. 08774502
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08774502 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08774502 Supplier Corning™ Supplier No. 354218
Only null left

Reliable and consistent dye used for detection of live cells and key functional activities in variety of cell-based assays

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Specifications

Content And Storage -20°C
For Use With (Application) Live Cell Detection, Cell Labeling
For Use With (Equipment) Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells
Quantity 100 mg

For research use only.