missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Collagen IV, Human
GREENER_CHOICE

Catalog No. CB40245
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB40245 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB40245 Supplier Corning™ Supplier No. 354245

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Specifications

Concentration 1 to 5 μg/cm2
For Use With (Application) Cell Attachment and Proliferation
Formulation Frozen in 100 mm borate buffer pH 8.0
pH Range 8
Quantity 0.25 mg
Source Human Placenta
Product Type Collagen, Human Type IV
Content And Storage -70°C