missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Clear Polypropylene 384-Well Deep-Well Plates

Catalog No. 361004520
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
361004520 Non-sterile
07202501 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361004520 Supplier Corning™ Supplier No. 3347

Ideal for high-throughput applications requiring added volume

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

Order Info

Packaged in bags of ten

Certifications

Certified DNase- and RNase-free

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Packaging Bags of 5
Color Clear
Product Type 384 Deep-Well Microplate
Material Polypropylene
Volume (Metric) Well 240 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
For Use With (Application) For high throughput applications requiring added volume
No. per Case 50/Cs.
No. per Pack 5/Pk.
Quantity 50/Cs.
Show More Show Less