missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CELLSTACK™ Culture Chambers
GREENER_CHOICE

Catalog No. p-300921
Change view
Click to view available options
Culture Area:
1272 cm²
25,440 cm²
3180 cm²
636 cm²
6360 cm²
Surface Treatment:
CellBIND
Tissue Culture-treated
Ultra-low Attachment
Quantity:
2/Cs.
5/Cs.
6/Cs.
8/Cs.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Surface Treatment Quantity
05539094 636 cm² Tissue Culture-treated 8/Cs.
05539096 6360 cm² Tissue Culture-treated 2/Cs.
05539098 3180 cm² Tissue Culture-treated 8/Cs.
08757226 636 cm² Ultra-low Attachment 8/Cs.
07201630 3180 cm² Tissue Culture-treated 2/Cs.
07201820 25,440 cm² Tissue Culture-treated 2/Cs.
0720191 6360 cm² CellBIND 6/Cs.
0720190 1272 cm² CellBIND 5/Cs.
0720195 3180 cm² CellBIND 2/Cs.
0720193 636 cm² CellBIND 8/Cs.
08757213 6360 cm² CellBIND 2/Cs.
05539097 6360 cm² Tissue Culture-treated 6/Cs.
05539095 1272 cm² Tissue Culture-treated 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 05539094 Supplier Corning™ Supplier No. 3268
Only null left

Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.

  • Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
  • Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
  • Sterilized by gamma irradiation
  • Leak tested

  • Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
  • Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation

Order Info

Individually packaged

Certifications

Certified nonpyrogenic.

Compliance

Polystyrene material meets USP Class VI requirements; provides superior optical clarity and structural integrity.

Specifications

Capacity (English) 4.22 to 6.42 oz.
Certifications/Compliance USP Class VI
Closure Color Orange
Diameter (Metric) Closure 33mm
Capacity (Metric) 125 to 190 mL
Material Polystyrene
Includes 1 Chamber
Product Type Culture Chamber
Graduated Yes
Culture Area 636 cm²
Surface Treatment Tissue Culture-treated
Quantity 8/Cs.
Sterilization Method Gamma Irradiated
Show More Show Less