Learn More
Reagent Grade Water Tested to USP Purified Water, Corning™
Catalog No. mt46002lf
Change view
Click to view available options
Quantity:
2 x 4 L
Catalog No. Quantity
MT46002LF 2 x 4 L
1 options
Catalog No. MT46002LF Supplier Corning Incorporated Supplier No. 46002LF
Only null left
Used in common laboratory applications requiring highly pure water. Corning™ Reagent Grade Water Tested to USP Purified Water is used in the preparation of reagents, rinsing glassware and plastic ware.
  • Tested to USP monograph for purified water
  • DNase, RNase, Protease-free
  • Endotoxin levels ≤0.025EU/mL
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Quantity 2 x 4 L