missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Tryptic Soy Broth, Animal-Free, Liquid

Catalog No. MT46060LB
Change view
Click to view available options
Quantity:
20,000 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT46060LB 20,000 mL
MT46060CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT46060LB Supplier Corning™ Supplier No. 46060LB
  • ONLINE EXCLUSIVE PRICE
    $1,758.05
    $1,484.56 / Each

    Save $273.49

    16.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For an accurate assessment of sterility throughout the manufacturing process

Tryptic Soy Broth is a sterile-filtered, ready-to-use solution for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious micro-organisms. This growth medium has animal-free components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.

Order Info

Shipped under ambient conditions

Specifications

Certifications/Compliance USP
Content And Storage 15°C to 30°C
Form Liquid
For Use With (Application) For testing the sterility of animal-free biopharmaceutical processes
Quantity 20,000 mL