missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sodium Bicarbonate Solution

Catalog No. MT25035CI
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25035CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25035CI Supplier Corning™ Supplier No. 25035CI
Only null left

Used as part of buffering system commonly used to maintain physiological pH 7.2 to 7.4 in a culture environment

Technical Advantage:  Product manufactured under industry-leading standards for low endotoxin content and superior results. Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7.2 - 7.4 in a culture environment.  Bicarbonate is a weak buffer with that makes maintaining a pH range of 7.2 - 7.6 somewhat difficult, but it is non-toxic, and is a nutrient source.  Sodium Bicarbonate #25-035 is offered in a 7.5% solution prepared in Water for Injection (WFI) quality water.  See the addition Chart included in the Technical Information section to the left for information on proper use. -  Complete Certificate of Analysis available for each lot, along with full formulation information

Corning media certificates are available on our Resource Library.

Specifications

Content And Storage 15° to 30°C
Form Liquid
Product Type Sodium Bicarbonate
Sterility Sterile-filtered
Format Plastic Bottle
pH 7.8 to 8.2
Quantity 6 x 100 mL