missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine

Catalog No. MT17305CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
Without Additives:
No Glutamine
No Glutamine, No Calcium, No Magnesium
No Glutamine, No Phenol Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Without Additives
MT17305CV 6 x 500 mL No Glutamine, No Phenol Red
MT15010CM 6 x 1 L No Glutamine
MT15010CV 6 x 500 mL No Glutamine
MT15015CV 6 x 500 mL No Glutamine, No Calcium, No Magnesium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MT17305CV Supplier Corning™ Supplier No. 17305CV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guaranteed low endotoxin levels ≤0.025EU/mL.

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.  This research indicated that additions made to BME aided cell propagation.  MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells.  When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayer, such as fibroblast.  This formulation contains sodium bicarbonate and does not contain L-glutamine.

Specifications

Form Liquid
Product Type MEM Eagle/EBSS
Sterility Sterile
With Additives Earle's Balanced Salt Solution (EBSS)
Without Additives No Glutamine, No Phenol Red
Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C