missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) with Glutamine

Catalog No. MT10010CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT10010CV 6 x 500 mL
MT10010CM 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT10010CV Supplier Corning™ Supplier No. 10010CV
  • ONLINE EXCLUSIVE PRICE
    $175.21
    $150.81 / Case of 6 Each

    Save $24.40

    14.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guaranteed low endotoxin levels ≤0.025EU/mL

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. This research indicated that additions made to BME aided cell propagation. MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.  This formulation contains Earle's salts and L-glutamine.

Specifications

Form Liquid
Product Type MEM Eagle/EBSS
Sterility Sterile
With Additives Glutamine, Earle's Balanced Salt Solution (EBSS)
Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C