Learn More
Corning™ Minimum Essential Medium Eagle (Mod.)
Catalog No. MT50011PC
Change view
Click to view available options
Quantity:
50 L
10 L
With Additives:
Earle's Balanced Salt Solution (EBSS)
Earle's Balanced Salt Solution (EBSS), Glutamine
Earle's Balanced Salt Solution (EBSS), Glutamine, Non Essential Amino Acids (NEAA)
Without Additives:
No Glutamine, No Sodium Bicarbonate
No Sodium Bicarbonate
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT50011PC 50 L Earle's Balanced Salt Solution (EBSS), Glutamine, Non Essential Amino Acids (NEAA) No Sodium Bicarbonate
MT50010PB 10 L Earle's Balanced Salt Solution (EBSS), Glutamine No Sodium Bicarbonate
MT50010PC 50 L Earle's Balanced Salt Solution (EBSS), Glutamine No Sodium Bicarbonate
MT50011PB 10 L Earle's Balanced Salt Solution (EBSS), Glutamine, Non Essential Amino Acids (NEAA) No Sodium Bicarbonate
MT51010PC 50 L Earle's Balanced Salt Solution (EBSS) No Glutamine, No Sodium Bicarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MT50011PC Supplier Corning™ Supplier No. 50011PC
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.  This research indicated that additions made to BME aided cell propagation.  MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells.  When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.  This formulation contains Earle's salts and L-glutamine, and does not contain sodium bicarbonate.
Specifications
With Additives Earle's Balanced Salt Solution (EBSS), Glutamine, Non Essential Amino Acids (NEAA)
Without Additives No Sodium Bicarbonate
Quantity 50 L
Content And Storage 2° to 8°C