missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Microbiological Media
Description
- For the maintenance and propagation of E. coli
- Terrific Broth supports higher cell densities than LB Broth, increasing plasmid yield
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Terrific Broth |
| Fill Volume | 6 x 1000 mL |
| Formulation | Casein Peptone 10g/L, NaCl 10g/L |
| For Use With (Application) | For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production |
| Packaging Type | Bottle |
| Quantity | 6 x 1000 mL |